2-(4-Biphenylyl)-4,6-dichloro-1,3,5-triazine ≥97.0% (by GC, total nitrogen)

Supplier: TCI America
Warning

B4940-1G B4940-200MG
TCB4940-1GEA 243 CAD
TCB4940-1G TCB4940-200MG
2-(4-Biphenylyl)-4,6-dichloro-1,3,5-triazine ≥97.0% (by GC, total nitrogen)
2-(4-Biphenylyl)-4,6-dichloro-1,3,5-triazine
Formula: C₁₅H₉Cl₂N₃
MDL Number: MFCD28556865
CAS Number: 10202-45-6

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (with Total Nitrogen) min. 97.0 %
Melting point 157.0 to 161.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR