7,7'-Bi[1,4]benzoxazino[2,3,4-kl]phenoxazine 98,0 HPLC_ASSAY_METHOD

Supplier: TCI America

Synonyms: HN-D1

B4908-1G B4908-200MG
TCB4908-1GEA 1167.62 CAD
TCB4908-1G TCB4908-200MG
7,7'-Bi[1,4]benzoxazino[2,3,4-kl]phenoxazine 98,0 HPLC_ASSAY_METHOD
7,7'-Bi[1,4]benzoxazino[2,3,4-kl]phenoxazine
Formula: C₃₆H₂₀N₂O₄
MW: 544.57 g/mol
Melting Pt: 376°C
CAS Number: 1395881-55-6

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 4.80 to 5.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR