2,7-Bis[N-(1-naphthyl)anilino]-9,9'-spirobi[9H-fluorene] ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

B4875-1G B4875-200MG
TCB4875-1GEA 439.71 CAD
TCB4875-1G TCB4875-200MG
2,7-Bis[N-(1-naphthyl)anilino]-9,9'-spirobi[9H-fluorene] ≥98.0% (by HPLC, total nitrogen)
2,7-Bis[N-(1-naphthyl)anilino]-9,9'-spirobi[9H-fluorene]
Formula: C₅₇H₃₈N₂
MW: 750.95 g/mol
MDL Number: MFCD12022428
CAS Number: 932739-76-9

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR