1-Bromo-3-n-octylbenzene ≥97.0% (by GC)

Supplier: TCI America
Warning

B4817-1G B4817-5G
TCB4817-1GEA 132.72 CAD
TCB4817-1G TCB4817-5G
1-Bromo-3-n-octylbenzene ≥97.0% (by GC)
1-Bromo-3-n-octylbenzene
Formula: C₁₄H₂₁Br
CAS Number: 663949-34-6

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR