3-Bromo-4-nitrobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B4786-1G B4786-5G
TCB4786-1GEA 112.18 CAD
TCB4786-1G TCB4786-5G
3-Bromo-4-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
3-Bromo-4-nitrobenzoic acid
Formula: C₇H₄BrNO₄
MW: 246.02
MDL Number: MFCD04117949
CAS Number: 101420-81-9

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 201.0 to 205.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR