2-Bromo-4'-methylpropiophenone ≥98.0% (by GC)

Supplier: TCI America
Warning

B4781-25G B4781-5G
TCB4781-25GEA 259.44 CAD
TCB4781-25G TCB4781-5G
2-Bromo-4'-methylpropiophenone ≥98.0% (by GC)
2-Bromo-4'-methylpropiophenone
Formula: C₁₀H₁₁BrO
Boiling Pt: 162°C
MDL Number: MFCD11131402
CAS Number: 1451-82-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 78.0 to 82.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR