2-Butoxyphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

B4700-1G
TCB4700-1GEA 85.95 CAD
TCB4700-1G
2-Butoxyphenylboronic acid (contains varying amounts of Anhydride)
2-Butoxyphenylboronic acid
Formula: C₁₀H₁₅BO₃
MDL Number: MFCD03427052
CAS Number: 91129-69-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 111.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR