Bis(trimethylsilylmethyl) sulfide ≥95.0% (by GC)

Supplier: TCI America
Warning

B4615-1ML
TCB4615-1MLEA 363.4 CAD
TCB4615-1ML
Bis(trimethylsilylmethyl) sulfide ≥95.0% (by GC)
Bis(trimethylsilylmethyl) sulfide
Formula: C₈H₂₂SSi₂
Boiling Pt: 85°C
Flash Pt: 65°C
MDL Number: MFCD00039797
CAS Number: 4712-51-0

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR