3-Bromofluoranthene 98,0 GC_ASSAY_METHOD

Supplier: TCI America
Warning

B4599-1G B4599-5G
TCB4599-1GEA 53.72 CAD
TCB4599-1G TCB4599-5G
3-Bromofluoranthene 98,0 GC_ASSAY_METHOD
3-Bromofluoranthene
Formula: C16H9Br
MW: 281.1467 g/mol
Boiling Pt: 207°C
MDL Number: MFCD07787460
CAS Number: 13438-50-1

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 106.0 to 200.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR