1,3-Bis(2-propynyloxy)benzene ≥97.0%

Supplier: TCI America

B4521-1G B4521-200MG
TCB4521-1GEA 663.6 CAD
TCB4521-1G TCB4521-200MG
1,3-Bis(2-propynyloxy)benzene ≥97.0%
1,3-Bis(2-propynyloxy)benzene

CAS Number: 26627-36-1
MDL Number: MFCD00224312
Molecular Formula: C12H10O2
Molecular Weight: 186.21
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 40
Lambda max.: 295 nm (H2O)Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: 1,3-Bis(propargyloxy)benzene

Formula: C₁₂H₁₀O₂
Melting Pt: 40 °C
MDL Number: MFCD00224312
CAS Number: 26627-36-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba318bb4b33015ed7f8989824cdd0f78a21e0572&vid=ba318bb4b33015ed7f8989824cdd0f78a21e0572&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR