1-Boc-3-cyanopiperidine ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: tert-butyl 3-cyanopiperidine-1-carboxylate, 1-Boc-3-cyanopiperidine, 3-Cyano-1-piperidinecarboxylic acid tert-Butyl ester, tert-Butyl 3-Cyano-1-piperidinecarboxylate, 1-Boc-3-piperidinecarbonitrile, 1-tert-Butoxycarbonyl-3-piperidinecarbonitrile, 1-tert-Butoxycarbonyl-3-cyanopiperidine

B4515-1G
TCB4515-1GEA 62.88 CAD
TCB4515-1G
1-Boc-3-cyanopiperidine ≥98.0% (by GC)
1-Boc-3-cyanopiperidine
Formula: C₁₁H₁₈N₂O₂
MW: 210.27 g/mol
Boiling Pt: 40°C
Storage Temperature: Refrigerator
MDL Number: MFCD01861222
CAS Number: 91419-53-3
UN: 3439
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 65.0 to 69.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR