2-Bromo-4-(trifluoromethyl)phenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B4492-1G B4492-5G
TCB4492-1GEA 121.66 CAD
TCB4492-1G TCB4492-5G
2-Bromo-4-(trifluoromethyl)phenol ≥98.0% (by GC, titration analysis)
2-Bromo-4-(trifluoromethyl)phenol
Formula: C₇H₄BrF₃O
MDL Number: MFCD06657954
CAS Number: 81107-97-3

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR