N-(tert-Butoxycarbonyl)propargylamine ≥98.0% (by GC)

Supplier: TCI America

B4472-1G B4472-5G
TCB4472-1GEA 35.38 CAD
TCB4472-1G TCB4472-5G
N-(tert-Butoxycarbonyl)propargylamine ≥98.0% (by GC)
N-(tert-Butoxycarbonyl)propargylamine

CAS Number: 92136-39-5
MDL Number: MFCD07367245
Molecular Formula: C8H13NO2
Molecular Weight: 155.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 170
Melting point (°C): 42
Flash Point (°C): 93
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry), Bioscience, Crosslinkers
Other Product Names / Synonyms: N-Boc-propargylamine, tert-Butyl 2-Propynylcarbamate, 2-Propynylcarbamic Acid tert-Butyl Ester

Formula: C₈H₁₃NO₂
Boiling Pt: 170 °C
Melting Pt: 42 °C
Flash Pt: 93 °C
MDL Number: MFCD07367245
CAS Number: 92136-39-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d9797c8a04d4c03d608e06b02f196fdd7d58bc5&vid=3d9797c8a04d4c03d608e06b02f196fdd7d58bc5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR