Bis(4-bromophenyl)sulfide ≥98.0% (by GC)

Supplier: TCI America

B4435-1G B4435-5G
TCB4435-1GEA 140.46 CAD
TCB4435-1G TCB4435-5G
Bis(4-bromophenyl)sulfide ≥98.0% (by GC)
Bis(4-bromophenyl)sulfide

CAS Number: 3393-78-0
Molecular Formula: C12H8Br2S
Molecular Weight: 344.06
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 210
Melting point (°C): 115

Formula: C₁₂H₈Br₂S
Boiling Pt: 210 °C
Melting Pt: 115 °C
MDL Number: MFCD00623633
CAS Number: 3393-78-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6583e58c7f90cc9059c5a78a7bac6da850073804&vid=6583e58c7f90cc9059c5a78a7bac6da850073804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR