(E)-1,2-Bis(2-naphthyl)-1,2-bis(1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-s-indacen-4-yl)disilene ≥97.0% (by HPLC)

Supplier: TCI America

Synonyms: (Eind)(2-Naph)Si=Si(2-Naph)(Eind)

B4422-50MG
TCB4422-50MGEA 641.32 CAD
TCB4422-50MG
(E)-1,2-Bis(2-naphthyl)-1,2-bis(1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-s-indacen-4-yl)disilene ≥97.0% (by HPLC)
(E)-1,2-Bis(2-naphthyl)-1,2-bis(1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-s-indacen-4-yl)disilene

CAS Number: 1253900-41-2
Molecular Formula: C76H104Si2
Molecular Weight: 1073.84
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Melting point (°C): 285
Lambda max.: 504 nm (THF)


Other Product Names / Synonyms: (Eind)(2-Naph)Si=Si(2-Naph)(Eind)

Formula: C₇₆H₁₀₄Si₂
MW: 1073.84 g/mol
Melting Pt: 285 °C
CAS Number: 1253900-41-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a1c87691346501c4507cfa92b6e4f781f2956de&vid=7a1c87691346501c4507cfa92b6e4f781f2956de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR