N-(tert-Butoxycarbonyl)-4-phenyl-L-phenylalanine ≥98.0%

Supplier: TCI America

Synonyms: N-Boc-4-phenyl-L-phenylalanine, Boc-Bip(4,4')-OH, Boc-4-phenyl-Phe-OH, Boc-Bip-OH

B4289-1G
TCB4289-1GEA 129.56 CAD
TCB4289-1G
N-(tert-Butoxycarbonyl)-4-phenyl-L-phenylalanine ≥98.0%
N-(tert-Butoxycarbonyl)-4-phenyl-L-phenylalanine

CAS Number: 147923-08-8
MDL Number: MFCD00191184
Molecular Formula: C20H23NO4
Molecular Weight: 341.41
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: 22 deg (C=2, MeOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-4-phenyl-L-phenylalanine, Boc-4-phenyl-Phe-OH, Boc-Bip-OH

Formula: C₂₀H₂₃NO₄
MDL Number: MFCD00191184
CAS Number: 147923-08-8

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR