Benzo[b]thiophene-3-carboxaldehyde ≥98.0% (by GC)

Supplier: TCI America

B4086-1G B4086-5G
TCB4086-1GEA 52.14 CAD
TCB4086-1G TCB4086-5G
Benzo[b]thiophene-3-carboxaldehyde ≥98.0% (by GC)
Benzo[b]thiophene-3-carboxaldehyde

CAS Number: 5381-20-4
MDL Number: MFCD00052376
Molecular Formula: C9H6OS
Molecular Weight: 162.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 133
Melting point (°C): 58
Flash Point (°C): 113
Lambda max.: 302 nm (EtOH)


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 3-Formylbenzo[b]thiophene, Thianaphthene-3-carboxaldehyde

Formula: C₉H₆OS
Boiling Pt: 133 °C
Melting Pt: 58 °C
Flash Pt: 113 °C
MDL Number: MFCD00052376
CAS Number: 5381-20-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74f5e0fb569c9acc7bb14ece2e6a94fa2af0a1f8&vid=74f5e0fb569c9acc7bb14ece2e6a94fa2af0a1f8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR