6-Bromo-1H-indazole ≥98.0% (by GC)

Supplier: TCI America

B4080-25G B4080-5G B4080-1G
TCB4080-25GEA 360.4 CAD
TCB4080-25G TCB4080-5G TCB4080-1G
6-Bromo-1H-indazole ≥98.0% (by GC)
6-Bromo-1H-indazole

CAS Number: 79762-54-2
MDL Number: MFCD03265457
Molecular Formula: C7H5BrN2
Molecular Weight: 197.04
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 183

Formula: C₇H₅BrN₂
Melting Pt: 183 °C
MDL Number: MFCD03265457
CAS Number: 79762-54-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e671c2995dea1f7d7668024a6326f8661d0e457&vid=0e671c2995dea1f7d7668024a6326f8661d0e457&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR