4-tert-Butyl-1-(3-sulfopropyl)pyridinium hydroxide Inner salt hydrate ≥98.0% (by HPLC, total nitrogen) for biochemical research

Supplier: TCI America

Synonyms: 3-(4-(tert-Butyl)pyridin-1-ium-1-yl)propane-1-sulfonate, 3-(4-tert.-Butyl-1-pyridinio)-1-propanesulphonate, 3-(4-(tert-Butyl)pyridin-1-ium-1-yl)propane-1-sulphonate, NDSB 256-4T, 3-(4-tert.-Butyl-1-pyridinio)-1-propanesulfonate

B4030-1G
TCB4030-1GEA 154.29 CAD
TCB4030-1G
4-tert-Butyl-1-(3-sulfopropyl)pyridinium hydroxide Inner salt hydrate ≥98.0% (by HPLC, total nitrogen) for biochemical research
4-tert-Butyl-1-(3-sulfopropyl)pyridinium hydroxide Inner salt hydrate

[for Biochemical Research]
CAS Number: 570412-84-9
MDL Number: MFCD06411435
Molecular Formula: C12H19NO3S
Molecular Weight: 257.35
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal


Product Categories: Bioscience, Non-detergent Sulfobetaines (NDSB)
Other Product Names / Synonyms: 3-(4-tert-Butyl-1-pyridinio)propanesulfonate, NDSB 256-4T

Formula: C₁₂H₁₉NO₃S
MW: 257.35 g/mol
MDL Number: MFCD06411435
CAS Number: 570412-84-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d27960b1978d719ead1fea1f07481cfef175ac06&vid=1844f5c31463daced18ba6027c91dd3552105edd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR