Bestatin hydrochloride ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride) ≥97.0% (by HPLC)

Supplier: TCI America

Synonyms: Bestatin hydrochloride, (S)-2-((2S,3R)-3-Amino-2-hydroxy-4-phenylbutanamido)-4-methylpentanoic acid hydrochloride, Bestatin HCl

B4000-25MG
TCB4000-25MGEA 289.3 CAD
TCB4000-25MG
Bestatin hydrochloride ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride) ≥97.0% (by HPLC)
Bestatin hydrochloride ([S-(R*,S*)]-N-(3-Amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride)

CAS Number: 65391-42-6
MDL Number: MFCD00058004
Molecular Formula: C16H24N2O4
Molecular Weight: 344.84
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Melting point (°C): 108
Specific rotation [a]20/D: -13 deg (C=0.5, 1mol/L HCl)
Storage Temperature: >-20°C


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Antitumor Ingredients
Other Product Names / Synonyms: Ubenimex Hydrochloride

Formula: C₁₆H₂₅ClN₂O₄
MW: 308.4 · 36.5 g/mol
Melting Pt: 108 °C
MDL Number: MFCD00058004
CAS Number: 65391-42-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2b6e45e2faee94c0b1f9f2d825f3ff9ed0e4383&vid=f2b6e45e2faee94c0b1f9f2d825f3ff9ed0e4383&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR