Benzo[b]thiophene-2-carboxaldehyde ≥98.0% (by GC)

Supplier: TCI America

B3777-5G
TCB3777-5GEA 131.14 CAD
TCB3777-5G
Benzo[b]thiophene-2-carboxaldehyde ≥98.0% (by GC)
Benzo[b]thiophene-2-carboxaldehyde

CAS Number: 3541-37-5
MDL Number: MFCD01075041
Molecular Formula: C9H6OS
Molecular Weight: 162.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 136
Melting point (°C): 35


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 2-Formylbenzo[b]thiophene

Formula: C₉H₆OS
Boiling Pt: 136 °C
Melting Pt: 35 °C
MDL Number: MFCD01075041
CAS Number: 3541-37-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a7a9d1b341a19fcf5a4456e50e41517b01511f6&vid=0a7a9d1b341a19fcf5a4456e50e41517b01511f6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR