2,2'-Bibenzo[d][1,3,2]dioxaborole ≥98.0% (by GC)

Supplier: TCI America

B3757-1G B3757-5G
TCB3757-1GEA 69.36 CAD
TCB3757-1G TCB3757-5G
2,2'-Bibenzo[d][1,3,2]dioxaborole ≥98.0% (by GC)
2,2'-Bibenzo[d][1,3,2]dioxaborole

CAS Number: 13826-27-2
Molecular Formula: C12H8B2O4
Molecular Weight: 237.81
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 196


Product Categories: Synthetic Organic Chemistry, Suzuki-Miyaura Cross-coupling Reaction, Boration
Other Product Names / Synonyms: 2,2'-Bi-1,3,2-benzodioxaborole

Formula: C₁₂H₈B₂O₄
Melting Pt: 196 °C
MDL Number: MFCD09842343
CAS Number: 13826-27-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=963d0bca89707cb0390610d593f2f5a3042791aa&vid=963d0bca89707cb0390610d593f2f5a3042791aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR