Bis(2-bromoethyl)ether ≥98.0%

Supplier: TCI America

Synonyms: 1-Bromo-2-(2-bromoethoxy)ethane, 2-Bromoethyl ether

B3752-25G B3752-5G
TCB3752-25GEA 143.78 CAD
TCB3752-25G TCB3752-5G
Bis(2-bromoethyl)ether ≥98.0%
Bis(2-bromoethyl) ether

CAS Number: 5414-19-7
MDL Number: MFCD00039196
Molecular Formula: C4H8Br2O
Molecular Weight: 231.92
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 45
Flash Point (°C): 85
Specific Gravity (20/20): 1.86


Other Product Names / Synonyms: 2,2'-Dibromodiethyl Ether, 2-Bromoethyl Ether

Formula: C₄H₈Br₂O
MW: 231.93 g/mol
Boiling Pt: 45 °C
Density: 1.86 (20 °C)
Flash Pt: 85 °C
MDL Number: MFCD00039196
CAS Number: 5414-19-7
UN: 2810
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5a1b4cdacaf19e8a7faaff8f43896b5d5002e9c&vid=f5a1b4cdacaf19e8a7faaff8f43896b5d5002e9c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR