4-Bromo-1H-pyrazole ≥98.0% (by GC)

Supplier: TCI America

B3745-25G B3745-5G
TCB3745-25GEA 137.46 CAD
TCB3745-25G TCB3745-5G
4-Bromo-1H-pyrazole ≥98.0% (by GC)
4-Bromo-1H-pyrazole

CAS Number: 2075-45-8
MDL Number: MFCD00075602
Molecular Formula: C3H3BrN2
Molecular Weight: 146.98
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 260
Melting point (°C): 97

Formula: C₃H₃BrN₂
Boiling Pt: 260 °C
Melting Pt: 97 °C
MDL Number: MFCD00075602
CAS Number: 2075-45-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ff81b418482d7c3c6241a4a13dc3bf410c89fbd&vid=9ff81b418482d7c3c6241a4a13dc3bf410c89fbd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR