tert-Butyl (4R,6R)-2-[6-[2-[2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)pyrrol-1-yl]ethyl]-2,2-dimethyl-1,3-dioxan-4-yl]acetate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: tert-Butyl 2-((4R,6R)-6-(2-(2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl)ethyl)-2,2-dimethyl-1,3-dioxan-4-yl)acetate, Atorvastatin Acetonide tert-Butyl Ester

B3690-1G B3690-5G
TCB3690-1GEA 116.92 CAD
TCB3690-1G TCB3690-5G
tert-Butyl (4R,6R)-2-[6-[2-[2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)pyrrol-1-yl]ethyl]-2,2-dimethyl-1,3-dioxan-4-yl]acetate ≥98.0% (by HPLC, total nitrogen)
tert-Butyl (4R,6R)-2-[6-[2-[2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)pyrrol-1-yl]ethyl]-2,2-dimethyl-1,3-dioxan-4-yl]acetate

CAS Number: 125971-95-1
MDL Number: MFCD04039904
Molecular Formula: C40H47FN2O5
Molecular Weight: 654.82
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 146
Specific rotation [a]20/D: 7 deg (C=1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Chiral Building Blocks
Other Product Names / Synonyms: Atorvastatin Acetonide tert-Butyl Ester

Formula: C₄₀H₄₇FN₂O₅
MW: 654.82 g/mol
Melting Pt: 146 °C
MDL Number: MFCD04039904
CAS Number: 125971-95-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8fb0577be947432d379f15a57fde25e0f47debbd&vid=8fb0577be947432d379f15a57fde25e0f47debbd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR