N-[2-(tert-Butoxycarbonylamino)ethyl]glycineethyl ester ≥95.0%

Supplier: TCI America

Synonyms: N-[2-(Boc-amino)ethyl]glycine Ethyl Ester, Ethyl 2-((2-((tert-butoxycarbonyl)amino)ethyl)amino)acetate

B3632-1G
TCB3632-1GEA 261.17 CAD
TCB3632-1G
N-[2-(tert-Butoxycarbonylamino)ethyl]glycineethyl ester ≥95.0%
N-[2-(tert-Butoxycarbonylamino)ethyl]glycineethyl ester

CAS Number: 72648-80-7
Molecular Formula: C11H22N2O4
Molecular Weight: 246.31
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid


Product Categories: Bioscience, Peptide Nucleic Acids and Related Reagents (PNA)
Other Product Names / Synonyms: N-[2-(Boc-amino)ethyl]glycine Ethyl Ester

Formula: C₁₁H₂₂N₂O₄
MDL Number: MFCD00672310
CAS Number: 72648-80-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7cf699803a51c994f6aa4430213f1752b9921a4&vid=e7cf699803a51c994f6aa4430213f1752b9921a4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR