N-(tert-Butoxycarbonyl)-3,4-difluoro-L-phenylalanine ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America

B3627-1G
TCB3627-1GEA 366.56 CAD
TCB3627-1G
N-(tert-Butoxycarbonyl)-3,4-difluoro-L-phenylalanine ≥95.0% (by HPLC, titration analysis)
N-(tert-Butoxycarbonyl)-3,4-difluoro-L-phenylalanine

CAS Number: 198474-90-7
MDL Number: MFCD00672518
Molecular Formula: C14H17F2NO4
Molecular Weight: 301.29
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Melting point (°C): 83
Specific rotation [a]20/D: 5 deg (C=1, MeOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-3,4-difluoro-L-phenylalanine, Boc-Phe(3,4-F)-OH

Formula: C₁₄H₁₇F₂NO₄
Melting Pt: 83 °C
MDL Number: MFCD00672518
CAS Number: 198474-90-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6a7ab5cc74b301fdda766ad136af2b3079fc4ec&vid=e6a7ab5cc74b301fdda766ad136af2b3079fc4ec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR