Bismuth(III) chloride, anhydrous ≥97.0%

Supplier: TCI America
Warning

Synonyms: Bismuth trichloride

B3546-250G B3546-25G
TCB3546-250GEA 308.1 CAD
TCB3546-250G TCB3546-25G
Bismuth(III) chloride, anhydrous ≥97.0%
Bismuth (III) chloride

CAS Number: 7787-60-2
MDL Number: MFCD00003461
Molecular Formula: BiCl3
Molecular Weight: 315.33
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Boiling point (°C): 447
Melting point (°C): 230

Formula: BiCl₃
Boiling Pt: 447 °C
Melting Pt: 230 °C
Density: 4.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003461
CAS Number: 7787-60-2
EINECS: 232-123-2
UN: 3260
ADR: 8,III
Merck Index: 13,01261

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR