4-Bromo-2,6-difluorobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3447-5G
TCB3447-5GEA 62.71 CAD
TCB3447-5G
4-Bromo-2,6-difluorobenzoic acid ≥98.0% (by GC, titration analysis)
4-Bromo-2,6-difluorobenzoic acid

CAS Number: 183065-68-1
MDL Number: MFCD03094085
Molecular Formula: C7H3O2BrF2
Molecular Weight: 237.00
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 201

Formula: C₇H₃BrF₂O₂
Melting Pt: 201 °C
MDL Number: MFCD03094085
CAS Number: 183065-68-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d31d5f862ab87d68e2a5d0dab6f4560ece9760d&vid=9d31d5f862ab87d68e2a5d0dab6f4560ece9760d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR