Bis(2-methyl-3-furyl)disulfide ≥98.0%

Supplier: TCI America

B3360-5G
TCB3360-5GEA 61.62 CAD
TCB3360-5G
Bis(2-methyl-3-furyl)disulfide ≥98.0%
Bis(2-methyl-3-furyl)disulfide

CAS Number: 28588-75-2
MDL Number: MFCD01074774
Molecular Formula: C10H10O2S2
Molecular Weight: 226.31
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 92
Flash Point (°C): 110
Specific Gravity (20/20): 1.22

Formula: C₁₀H₁₀O₂S₂
Boiling Pt: 92 °C
Density: 1.22 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD01074774
CAS Number: 28588-75-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e4a227ec9d268d7a25dde5bc565973e632998c1&vid=0e4a227ec9d268d7a25dde5bc565973e632998c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR