O-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-serine ≥98.0%

Supplier: TCI America

Synonyms: N-Fmoc-O-tert-butyl-L-serine, N-a-Fmoc-O-t-butyl-L-serine, Fmoc-Ser(tBu)-OH, O-tert-Butyl-N-Fmoc-L-serine

B3168-25G B3168-5G
TCB3168-25GEA 137.14 CAD
TCB3168-25G TCB3168-5G
O-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-serine ≥98.0%
O-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-serine

CAS Number: 71989-33-8
MDL Number: MFCD00037127
Molecular Formula: C22H25NO5
Molecular Weight: 383.44
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 136
Specific rotation [a]20/D: 24 deg (C=1, EtOAc)


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: O-tert-Butyl-N-Fmoc-L-serine, Fmoc-Ser(tBu)-OH

Formula: C₂₂H₂₅NO₅
MW: 383.44 g/mol
Melting Pt: 136 °C
MDL Number: MFCD00037127
CAS Number: 71989-33-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2371e4080a2eb1541261831ceb33bd6aa6d80cc4&vid=2371e4080a2eb1541261831ceb33bd6aa6d80cc4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR