1,3-Bis(2,6-diisopropylphenyl)imidazolinium chloride ≥96.0% (by HPLC, total nitrogen)

Supplier: TCI America

B3157-5G B3157-500MG B3157-1G
TCB3157-5GEA 347.6 CAD
TCB3157-5G TCB3157-500MG TCB3157-1G
1,3-Bis(2,6-diisopropylphenyl)imidazolinium chloride ≥96.0% (by HPLC, total nitrogen)
1,3-Bis(2,6-diisopropylphenyl)imidazolinium chloride

CAS Number: 258278-25-0
MDL Number: MFCD07369796
Molecular Formula: C27H39ClN2
Molecular Weight: 427.07
Purity/Analysis Method: >96.0% (HPLC,N)
Form: Crystal
Melting point (°C): 293


Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Ligands
Other Product Names / Synonyms: SIPr.HCl

Formula: C₂₇H₃₉ClN₂
Melting Pt: 293 °C
MDL Number: MFCD07369796
CAS Number: 258278-25-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbed379ce1f6fbfd91197bdea76e7b757c430f04&vid=dbed379ce1f6fbfd91197bdea76e7b757c430f04&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR