2-Bromo-5-(trifluoromethoxy)phenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3108-25G B3108-5G
TCB3108-25GEA 273.34 CAD
TCB3108-25G TCB3108-5G
2-Bromo-5-(trifluoromethoxy)phenol ≥98.0% (by GC, titration analysis)
2-Bromo-5-(trifluoromethoxy)phenol

CAS Number: 205371-26-2
MDL Number: MFCD06660162
Molecular Formula: C7H4BrF3O2
Molecular Weight: 257.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 1.74


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Bromo-2-hydroxy-4-(trifluoromethoxy)benzene

Formula: C₇H₄BrF₃O₂
Density: 1.74 (20 °C)
MDL Number: MFCD06660162
CAS Number: 205371-26-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0aa159424d8d1b80b43c093b2ea6270e984e17e6&vid=0aa159424d8d1b80b43c093b2ea6270e984e17e6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR