Bis(2-aminophenyl)sulfide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2,2'-Diaminodiphenyl sulfide, 2-Aminophenyl sulfide, 2,2'-Thiodianiline, 2,2'-Diaminodiphenyl sulphide, 2-Aminophenyl Sulphide

B2928-25G
TCB2928-25GEA 230.68 CAD
TCB2928-25G
Bis(2-aminophenyl)sulfide ≥98.0% (by HPLC, titration analysis)
Bis(2-aminophenyl)sulfide

CAS Number: 5873-51-8
MDL Number: MFCD03094662
Molecular Formula: C12H12N2S
Molecular Weight: 216.30
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 87


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: 2-Aminophenyl Sulfide, 2,2'-Diaminodiphenyl Sulfide, 2,2'-Thiodianiline

Formula: C₁₂H₁₂N₂S
MW: 216.30 g/mol
Melting Pt: 87 °C
MDL Number: MFCD03094662
CAS Number: 5873-51-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f2de7ff12fd887ed0bbe723458835b804b00904&vid=9f2de7ff12fd887ed0bbe723458835b804b00904&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR