Benzo[b]thiophene-2-boronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

B2893-1G B2893-5G B2893-25G
TCB2893-1GEA 42.66 CAD
TCB2893-1G TCB2893-5G TCB2893-25G
Benzo[b]thiophene-2-boronic acid (contains varying amounts of Anhydride)
Benzo[b]thiophene-2-boronic acid

CAS Number: 98437-23-1
MDL Number: MFCD01075674
Molecular Formula: C8H7BO2S
Molecular Weight: 178.01
Form: Crystal
Color: White
Melting point (°C): 260

Formula: C₈H₇BO₂S
Melting Pt: 260 °C
MDL Number: MFCD01075674
CAS Number: 98437-23-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6cf5171597f186834ced4e6c0b36fe3fb3dca424&vid=6cf5171597f186834ced4e6c0b36fe3fb3dca424&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR