4'-Bromobiphenyl-4-ylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 4'-Bromo-4-biphenylboronic acid

B2860-1G
TCB2860-1GEA 295.46 CAD
TCB2860-1G
4'-Bromobiphenyl-4-ylboronic acid (contains varying amounts of Anhydride)
4'-Bromobiphenyl-4-ylboronic acid

CAS Number: 480996-05-2
MDL Number: MFCD02251652
Molecular Formula: C12H10BBrO2
Molecular Weight: 276.92
Form: Crystal
Color: White


Other Product Names / Synonyms: 4-(4-Bromophenyl)phenylboronic Acid, 4-(4-Bromophenyl)benzeneboronic Acid

Formula: C₁₂H₁₀BBrO₂
MW: 276.9216 g/mol
MDL Number: MFCD02251652
CAS Number: 480996-05-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3bb727b5953b05d000495399abd6cd3254c4d000&vid=3bb727b5953b05d000495399abd6cd3254c4d000&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR