tert-Butyl (diethylphosphono)acetate ≥97.0%

Supplier: TCI America

Synonyms: tert-Butyl 2-(diethoxyphosphoryl)acetate, Diethyl(2-tert-butoxycarbonylmethyl)phosphonate

B2814-1G B2814-5G
TCB2814-1GEA 41.08 CAD
TCB2814-1G TCB2814-5G
tert-Butyl (diethylphosphono)acetate ≥97.0%
tert-Butyl (diethylphosphono)acetate

CAS Number: 27784-76-5
Molecular Formula: C10H21O5P
Molecular Weight: 252.25
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 152
Flash Point (°C): 109
Specific Gravity (20/20): 1.07


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: Diethyl (Boc-methyl)phosphonate, Diethyl (tert-Butoxycarbonylmethyl)phosphonate, Diethylphosphonoacetic Acid tert-Butyl Ester

Formula: C₁₀H₂₁O₅P
MW: 252.25 g/mol
Boiling Pt: 152 °C
Density: 1.07 (20 °C)
Flash Pt: 109 °C
MDL Number: MFCD00075414
CAS Number: 27784-76-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea5fb6ec8766dcde1519032f136581a40108093&vid=bea5fb6ec8766dcde1519032f136581a40108093&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR