3-Bromo-4-chlorobenzotrifluoride ≥97.0%

Supplier: TCI America

B2743-25G
TCB2743-25GEA 82.63 CAD
TCB2743-25G
3-Bromo-4-chlorobenzotrifluoride ≥97.0%
3-Bromo-4-chlorobenzotrifluoride

CAS Number: 454-78-4
Molecular Formula: C7H3BrClF3
Molecular Weight: 259.45
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 190
Melting point (°C): -22
Flash Point (°C): 94
Specific Gravity (20/20): 1.75


Other Product Names / Synonyms: 2-Bromo-1-chloro-4-(trifluoromethyl)benzene, 3-Bromo-4-chloro-alpha,alpha,alpha-trifluorotoluene

Formula: C₇H₃BrClF₃
Boiling Pt: 190 °C
Melting Pt: –22 °C
Density: 1.75 (20 °C)
Flash Pt: 94 °C
MDL Number: MFCD00018093
CAS Number: 454-78-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc4107252baa223db41d85b462760fecc20212fe&vid=bc4107252baa223db41d85b462760fecc20212fe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR