1,3-Bis(2-hydroxyethylsulfonyl)-2-propanol ≥93.0%

Supplier: TCI America

B2699-25G
TCB2699-25GEA 113.76 CAD
TCB2699-25G
1,3-Bis(2-hydroxyethylsulfonyl)-2-propanol ≥93.0%
1,3-Bis(2-hydroxyethylsulfonyl)-2-propanol

CAS Number: 67006-34-2
Molecular Formula: C7H16O7S2
Molecular Weight: 276.32
Purity/Analysis Method: >93.0% (GC)
Form: Crystal

Formula: C₇H₁₆O₇S₂
CAS Number: 67006-34-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26c931304e4bf482c9676f0b98a3a9b26d3cd9d8&vid=26c931304e4bf482c9676f0b98a3a9b26d3cd9d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR