9,9-Bis(4-amino-3-methylphenyl)fluorene ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B2693-5G B2693-25G
TCB2693-5GEA 118.5 CAD
TCB2693-5G TCB2693-25G
9,9-Bis(4-amino-3-methylphenyl)fluorene ≥98.0% (by titrimetric analysis)
9,9-Bis(4-amino-3-methylphenyl)fluorene

CAS Number: 107934-60-1
Molecular Formula: C27H24N2
Molecular Weight: 376.50
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 233


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: 2,2'-Dimethyl-4,4'-(9-fluorenylidene)dianiline, 4,4'-(9-Fluorenylidene)di-o-toluidine

Formula: C₂₇H₂₄N₂
Melting Pt: 233 °C
MDL Number: MFCD04038168
CAS Number: 107934-60-1

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR