(1S,2S)-1,2-Bis(2,4,6-trimethylphenyl)ethylenediamine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (1S,2S)-1,2-Diamino-1,2-dimesitylethane, (1S,2S)-1,2-Dimesitylethylenediamine

B2317-500MG B2317-100MG
TCB2317-500MGEA 593.45 CAD
TCB2317-500MG TCB2317-100MG
(1S,2S)-1,2-Bis(2,4,6-trimethylphenyl)ethylenediamine ≥97.0% (by GC, titration analysis)
(1S,2S)-1,2-Bis(2,4,6-trimethylphenyl)ethylenediamine

CAS Number: 186769-18-6
MDL Number: MFCD06797064
Molecular Formula: C20H28N2
Molecular Weight: 296.46
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 85
Specific rotation [a]20/D: -202 deg (C=1, CH2Cl2)


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands, Chiral Building Blocks
Other Product Names / Synonyms: (1S,2S)-1,2-Diamino-1,2-dimesitylethane, (1S,2S)-1,2-Dimesitylethylenediamine

Formula: C₂₀H₂₈N₂
MW: 296.46 g/mol
Melting Pt: 85 °C
MDL Number: MFCD06797064
CAS Number: 186769-18-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d3eb5032b09a81c3979abdf3e29d9120b046802&vid=8d3eb5032b09a81c3979abdf3e29d9120b046802&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR