N,N-Bis(2-hydroxyethyl)-p-toluenesulfonamide ≥97.0% (by HPLC)

Supplier: TCI America

Synonyms: N,N-Bis(2-hydroxyethyl)-4-methylbenzenesulfonamide, N,N-Bis(2-hydroxyethyl)-4-methylbenzenesulphonamide

B2263-25G
TCB2263-25GEA 222.78 CAD
TCB2263-25G
N,N-Bis(2-hydroxyethyl)-p-toluenesulfonamide ≥97.0% (by HPLC)
N,N-Bis(2-hydroxyethyl)-p-toluenesulfonamide

CAS Number: 7146-67-0
MDL Number: MFCD00020582
Molecular Formula: C11H17NO4S
Molecular Weight: 259.32
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Melting point (°C): 100

Formula: C₁₁H₁₇NO₄S
MW: 259.32 g/mol
Melting Pt: 100 °C
MDL Number: MFCD00020582
CAS Number: 7146-67-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6829e218c694ccb388766afd45fa7fe9b98c608&vid=a6829e218c694ccb388766afd45fa7fe9b98c608&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR