Boc-b-Alanine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B2176-25G B2176-5G
TCB2176-25GEA 77.15 CAD
TCB2176-25G TCB2176-5G
Boc-b-Alanine ≥98.0% (by HPLC, titration analysis)
Boc-b-Alanine

CAS Number: 3303-84-2
MDL Number: MFCD00037291
Molecular Formula: C8H15NO4
Molecular Weight: 189.21
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 77


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-beta-alanine, Boc-beta-Ala-OH

Formula: C₈H₁₅NO₄
Melting Pt: 77 °C
MDL Number: MFCD00037291
CAS Number: 3303-84-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e666699a6c94b90a6a152a7c01c81f2945461ce&vid=8e666699a6c94b90a6a152a7c01c81f2945461ce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR