Tridecyl borate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B2096-5G
TCB2096-5GEA 303.36 CAD
TCB2096-5G
Tridecyl borate ≥98.0% (by titrimetric analysis)
Tridecyl borate

CAS Number: 20236-81-1
MDL Number: MFCD00026531
Molecular Formula: C30H63BO3
Molecular Weight: 482.64
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 258
Specific Gravity (20/20): 0.85


Other Product Names / Synonyms: Boric Acid Tridecyl Ester

Formula: C₃₀H₆₃BO₃
Boiling Pt: 258 °C
Density: 0.85 (20 °C)
MDL Number: MFCD00026531
CAS Number: 20236-81-1

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR