3,5-Bis(tert-butyldiphenylsilyloxy)benzyl alcohol ≥95.0%

Supplier: TCI America

B2052-1G
TCB2052-001GEA 246.32 CAD
TCB2052-001G
3,5-Bis(tert-butyldiphenylsilyloxy)benzyl alcohol ≥95.0%
3,5-Bis(tert-butyldiphenylsilyloxy)benzyl alcohol

CAS Number: 182250-70-0
MDL Number: MFCD02093440
Molecular Formula: C39H44O3Si2
Molecular Weight: 616.95
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 108

Formula: C₃₉H₄₄O₃Si₂
Melting Pt: 108 °C
MDL Number: MFCD02093440
CAS Number: 182250-70-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7dba2ce4da03a188f9d72a966bef169e6f4d0508&vid=7dba2ce4da03a188f9d72a966bef169e6f4d0508&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR