2-Bromothioanisole ≥96.0%

Supplier: TCI America
Warning

B2023-5G B2023-25G
TCB2023-005GEA 75.84 CAD
TCB2023-005G TCB2023-025G
2-Bromothioanisole ≥96.0%
2-Bromothioanisole

CAS Number: 19614-16-5
MDL Number: MFCD00000066
Molecular Formula: C7H7BrS
Molecular Weight: 203.10
Purity/Analysis Method: >96.0% (GC)
Form: Liquid
Boiling point (°C): 256
Flash Point (°C): 110
Specific Gravity (20/20): 1.54


Other Product Names / Synonyms: 1-Bromo-2-(methylthio)benzene, 2-Bromophenyl Methyl Sulfide

Formula: C₇H₇BrS
Boiling Pt: 256 °C
Melting Pt: –24.5 °C
Density: 1.54 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00000066
CAS Number: 19614-16-5
EINECS: 243-183-4
UN: 3334
ADR: 9,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=596dfd4d85f3fae2c06bd54224efb28933d1d586&vid=596dfd4d85f3fae2c06bd54224efb28933d1d586&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR