(2-Bromophenyl)acetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-(2-Bromophenyl)acetic acid

B1957-5G B1957-25G
TCB1957-005GEA 45.82 CAD
TCB1957-005G TCB1957-025G
(2-Bromophenyl)acetic acid ≥98.0% (by GC, titration analysis)
(2-Bromophenyl)acetic acid

CAS Number: 18698-97-0
MDL Number: MFCD00004314
Molecular Formula: C8H7BrO2
Molecular Weight: 215.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 106

Formula: (C₆H₄Br)CH₂COOH
MW: 215.05 g/mol
Boiling Pt: 308
Melting Pt: 106 °C
MDL Number: MFCD00004314
CAS Number: 18698-97-0
EINECS: 242-509-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02e1b5001b74bae93edcc3ba5757dd6946b1fae5&vid=02e1b5001b74bae93edcc3ba5757dd6946b1fae5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR