(R)-tert-Butyl(1-benzylpyrrolidin-3-yl)carbamate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (3R)-(+)-1-Benzyl-3-(tert-butoxycarbonylamino)pyrrolidine, (R)-tert-Butyl (1-benzylpyrrolidin-3-yl)carbamate

B1932-5G
TCB1932-005GEA 142.2 CAD
TCB1932-005G
(R)-tert-Butyl(1-benzylpyrrolidin-3-yl)carbamate ≥98.0% (by GC, titration analysis)
(R)-tert-Butyl(1-benzylpyrrolidin-3-yl)carbamate

CAS Number: 131878-23-4
MDL Number: MFCD01321148
Molecular Formula: C16H24N2O2
Molecular Weight: 276.38
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 80
Specific rotation [a]20/D: 18.5 deg (C=2, EtOH)

Formula: C₁₆H₂₄N₂O₂
MW: 276.3740 g/mol
Melting Pt: 80 °C
MDL Number: MFCD01321148
CAS Number: 131878-23-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aec277a5906f303aca289b0e2bcfb2c05ac5dc74&vid=aec277a5906f303aca289b0e2bcfb2c05ac5dc74&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR