4-Bromo-3-methylanisole ≥97.0%

Supplier: TCI America

B1782-250G B1782-25G
TCB1782-250GEA 952.74 CAD
TCB1782-250G TCB1782-025G
4-Bromo-3-methylanisole ≥97.0%
4-Bromo-3-methylanisole

CAS Number: 27060-75-9
MDL Number: MFCD00060687
Molecular Formula: C8H9BrO
Molecular Weight: 201.06
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 135
Flash Point (°C): 110
Specific Gravity (20/20): 1.44


Other Product Names / Synonyms: 4-Bromo-3-methylanisole

Formula: C₈H₉BrO
Boiling Pt: 135 °C
Density: 1.44 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00060687
CAS Number: 27060-75-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8c3d2b32d7028533536fab397f86f959a804616&vid=d8c3d2b32d7028533536fab397f86f959a804616&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR