(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP) ≥98.0% (by HPLC)

Supplier: TCI America

B1774-5G B1774-25G
TCB1774-005GEA 130.37 CAD
TCB1774-005G TCB1774-25G
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP) ≥98.0% (by HPLC)
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)

[Coupling Reagent for Peptide]
CAS Number: 128625-52-5
MDL Number: MFCD00077411
Molecular Formula: C18H28F6N6OP2
Molecular Weight: 520.40
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Storage Temperature: 0-10°C

Formula: C₁₈H₂₈F₆N₆OP₂
MDL Number: MFCD00077411
CAS Number: 128625-52-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49bbe8235daec3d77e154273e9d971f18c35045a&vid=ee7fcaf88ca32c1807accbbc31e3b8f7254dfe6e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR